Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis B Virus Mouse anti-Avian, Human, Clone: C1-5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB16988
Description
Anti-Hepatitis B Virus Antibody, core antigen, clone C1-5, a.a. 74-89 is an antibody against Hepatitis B Virus for use in ELISA, immunofluoresence, immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry.
Specifications
| Hepatitis B Virus | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 100 μL | |
| Avian, Human | |
| Ascites |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| C1-5 | |
| Ascitic fluid. Liquid in DMEM, containing 0.1% sodium azide. | |
| Recombinant purified HBcAg from E.coli | |
| Primary | |
| −20°C for up to 12 months in undiluted aliquots. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction