Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus Surface Antigen Mouse anti-Human, Clone: H21F8-1, MilliporeSigma™

Catalog No. MAB10234MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10234MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10234MI Supplier MilliporeSigma Supplier No. MAB10234
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Hepatitis B Virus Surface Antigen Antibody, clone H21F8-1 detects level of Hepatitis B Virus Surface Antigen and has been published and validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Virus Surface Antigen
Applications Immunofluorescence
Classification Monoclonal
Clone H21F8-1
Conjugate Unconjugated
Formulation Liquid ascites fluid
Host Species Mouse
Immunogen Hepatitis B virus surface antigen
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for up to 12 months.
Form Ascites
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.