Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus Surface Antigen Mouse anti-Human, Clone: H25B10, MilliporeSigma™

Catalog No. MAB10201MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10201MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10201MI Supplier MilliporeSigma Supplier No. MAB10201
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Hepatitis B Virus Surface Antigen using this Anti-Hepatitis B Virus Surface Antigen Antibody, clone H25B10 validated for use in ELISA and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis B Virus Surface Antigen
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone H25B10
Conjugate Unconjugated
Formulation Liquid ascites fluid containing no preservatives.
Host Species Mouse
Immunogen Hepatitis B virus surface antigen
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for one year
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.