Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis E Virus Mouse anti-Human, Clone: 1E6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8002
Description
Anti-Hepatitis E Virus Antibody, aa 434-457, clone 1E6 detects level of Hepatitis E Virus and has been published and validated for use in western blot, ELISA and immunofluoresence.
Specifications
Hepatitis E Virus | |
Monoclonal | |
Unconjugated | |
Mouse | |
Primary | |
2-8°C for one year | |
IgG2b |
ELISA, Immunofluorescence, Western Blot | |
1E6 | |
Liquid in 0.02M PB with 0.25M NaCl, pH 7.6. Contains 0.1% sodium azide. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction