Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis E Virus Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB8305MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB8305MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB8305MI Supplier MilliporeSigma Supplier No. AB8305
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Hepatitis E Virus using this Anti-Hepatitis E Virus Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis E Virus
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified. Liquid in PBS with 0.02% sodium azide.
Host Species Rabbit
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquotes for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.