Learn More
HER-2/neu Mouse anti-Human, FITC, Clone: NEU 24.7, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 340553
Description
Anti-HER-2/neu recognizes a single protein of 185kDa from HER-2 transfected cells.The HER-2/neu antigen, product of the c-erbB2 gene, is a surface marker on some breast cancer cells. Amplification and/or overexpression of HER-2/neu occurs in multiple human malignancies. Overexpression is reported to be strongly correlated with progression of human ovarian and breast carcinomas. Anti-HER-2/neu stains the external domain of this protein on cells such as the strongly expressing line SKBR3 and the weakly expressing line MCF7.
Applications:
Flow Cytometry

Specifications
HER-2/neu | |
Monoclonal | |
FITC | |
Mouse | |
50 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
NEU 24.7 | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
SKBR3 human breast carcinoma cells | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.