Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hect domain and RLD 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018628 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-628 Supplier Abnova Corporation Supplier No. H00008916D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human HERC3 protein.

HERC3 belongs to the HERC family of ubiquitin ligases, all of which contain a HECT domain and at least 1 RCC1 (MIM 179710)-like domain (RLD). The 350-amino acid HECT domain is predicted to catalyze the formation of a thioester with ubiquitin before transferring it to a substrate, and the RLD is predicted to act as a guanine nucleotide exchange factor for small G proteins (Hochrainer et al., 2005 [PubMed 15676274]).[supplied by OMIM

Sequence: MLCWGYWSLGQPGISTNLQGIVAEPQVCGFISDRSVKEVACGGNHSVFLLEDGEVYTCGLNTKGQLGHEREGNKPEQIGALADQHIIHVACGESHSLALSDRGQLFSWGAGSDGQLGLMTTEDSVAVPRLIQKLNQQTILQVSCGNWHCLALAADGQFFTWGKNSHGQLGLGKEFPSQASPQRVRSLEGIPLAQVAAGGAHSFALSLSGAVFGWGMNNAGQLGLSDEKDRESPCHVKLLRTQKVVYISCGEEHTAVLTKSGGVFTFGAGSCGQLGHDSMNDEVNPRRVLELMGSEVTQIACGRQHTLAFVPSSGLIYAFGCGARGQLGTGHTCNVKCPSPVKGYWAAHSGQLSARADRFKYHIVKQIFSGGDQTFVLCSKYENYSPAVDFRTMNQAHYTSLINDETIAVWRQKLSEHNNANTINGVVQILSSAACWNGSFLEKKIDEHFKTSPKIPGIDLNSTRVLFEKLMNSQHSMILEQILNSFESCLIPQLSSSPPDVEAMRIYLILPEFPLLQDSKYYITLTIPLAMAILRLDTNPSKVLDNWWSQVCPKYFMKLVNLYKGAVLYLLRGRKTFLIPVLFNNYITAALKLLEKLYKVNLKVKHVEYDTFYIPEISNLVDIQEDYLMWFLHQAGMKARPSIIQDTVTLCSYPFIFDAQAKTKMLQTDAELQMQVAVNGANLQNVFMLLTLEPLLARSPFLVLHVRRNNLVGDALRELSIHSDIDLKKPLKVIFDGEEAVDAGGVTKEFFLLLLKELLNPIYGMFTYYQDSNLLWFSDTCFVEHNWFHLIGITCGLAIYNSTVVDLHFPLALYKKLLNVKPGLEDLKELSPTEGRSLQELLDYPGEDVEETFCLNFTICRESYGVIEQKKLIPGGDNVTVCKDNRQEFVDAYVNYVFQISVHEWYTAFSSGFLKVCGGKVLELFQPSELRAMMVGNSNYNWEELEETAIYKGDYSATHPTVKLFWETFHEFPLEKKKKFLLFLTGSDRIPIYGMASLQIVIQSTASGEEYLPVAHTCYNLLDLPKYSSKEILSARLTQALDNYEGFSLA

Specifications

Antigen hect domain and RLD 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human HERC3 protein.
Formulation PBS with no preservative; pH 7.4
Gene HERC3
Gene Accession No. NM_014606.1
Gene Alias KIAA0032
Gene Symbols HERC3
Host Species Rabbit
Immunogen HERC3 (NP_055421.1, 1 a.a. ∼ 1050 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Ubiquitin/Proteasome
Primary or Secondary Primary
Gene ID (Entrez) 8916
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.