Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Virus 6 Mouse anti-Human, MilliporeSigma™

Catalog No. MAB8537MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8537MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8537MI Supplier MilliporeSigma Supplier No. MAB8537
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Herpes Virus 6 using this Anti-Herpes Virus 6 Antibody, A and B, gp 60/110 validated for use in immunofluoresence, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Herpes Virus 6
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.01Mphosphate buffered saline (PBS) pH 7.2, containing 0.1% sodium azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.