Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HES-1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5702
Description
This Anti-HES-1 Antibody is validated for use in immunohistochemistry, western blotting for the detection of HES-1.
Specifications
HES-1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing a solution of 50% saturated ammonium sulfate and PBS containing no preservatives. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse |
Immunohistochemistry, Western Blot | |
Unconjugated | |
HES1; Hes1; HL; HHL; FLJ20408; HRY; HES-1 | |
Synthetic peptide. | |
100 μg | |
NM_005524.2 | |
−80°C for up to 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction