Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HESCA-1 Mouse anti-Human, Clone: 051007-4A5, MilliporeSigma™

Catalog No. MAB4407MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4407MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4407MI Supplier MilliporeSigma Supplier No. MAB4407
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect HESCA-1 (Human Embryonic Stem Cell Antigen-1) using this Anti-HESCA-1 Antibody, clone 051007-4A5 validated for use in immunocytochemistry, immunohistochemistry, immunofluoresence, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HESCA-1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 051007-4A5
Conjugate Unconjugated
Formulation Liquid in 0.02M PBS, 0.25 NaCl, pH 7.6 with 0.1% Sodium Azide
Gene Symbols HESCA-1
Host Species Mouse
Immunogen BG01v hES cell line
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.