Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HESCA-1 Mouse anti-Human, FITC, Clone: 051007-4A5, MilliporeSigma™

Catalog No. FCMAB111FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB111FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB111FMI Supplier MilliporeSigma Supplier No. FCMAB111F

Mouse Monoclonal Antibody

Milli-Mark™ Anti-HESCA-1-FITC Antibody, clone 051007-4A5 detects level of HESCA-1 and has been published and validated for use in flow cytometry, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HESCA-1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone 051007-4A5
Conjugate FITC
Formulation Purified mouse monoclonal IgM conjugated to FITC in PBS with less than 0.09% sodium azide and 15mg/mL BSA.
Gene Symbols HESCA-1
Host Species Mouse
Immunogen BG01v hES cell line.
Purification Method G-25 size exclusion
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.