Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HET/SAF-B Mouse anti-Human, Clone: 6F7, MilliporeSigma™

Catalog No. 05588MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-588-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-588-MI Supplier MilliporeSigma Supplier No. 05588
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HET/SAF-B Antibody, clone 6F7 is a high quality Mouse Monoclonal Antibody for the detection of HET/SAF-B and has been validated in immunoprecipitation, western blot, immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HET/SAF-B
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6F7
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_002555
Gene Symbols SAFB, HET, DKFZp779C1727, HAP, SAF-B, SAFB1
Host Species Mouse
Immunogen Peptide corresponding to residues 345-357 of human HET/SAF-B
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.