Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heterochromatin Protein-1 Β Mouse anti-Human, Mouse, Clone: 1MOD-1A9, MilliporeSigma™

Catalog No. MAB3448MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3448MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3448MI Supplier MilliporeSigma Supplier No. MAB3448
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-Heterochromatin Protein-1 β Antibody, clone 1MOD-1A9 (Mouse Monoclonal Antibody) validated in ELISA, immunocytochemistry, immunohistochemistry, immunoprecipitation, western blotting to detect Heterochromatin Protein-1 β also known as HP1b.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Heterochromatin Protein-1 β
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1MOD-1A9
Conjugate Unconjugated
Formulation Liquid. Contains no preservative.
Gene Accession No. NM_007622.3
Gene Symbols CBX1, HP1-BETA, HP1B
Host Species Mouse
Immunogen Recombinant mouse HP1beta.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.