Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Hexahistidine (6HIS), Polyclonal, MP Biomedicals

Catalog No. MP0859269 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0859269 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0859269 Supplier MP Biomedicals, Inc Supplier No. 0859269

Host Species: Chicken

ELISA, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hexahistidine (6HIS)
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Chicken
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.