Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hexim 1 Mouse anti-Human, Clone: 10D2.1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE337
Description
This Anti-Hexim 1 Antibody, clone 10D2.1 is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of Hexim 1.
Specifications
Hexim 1 | |
Monoclonal | |
Unconjugated | |
NP_006451 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG2b κ |
Immunohistochemistry (Paraffin), Western Blot | |
10D2.1 | |
Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HEXIM1, CLP1, EDG1, HIS1, MAQ1 | |
GST-tagged recombinant protein corresponding to the C-terminus of human Hexim 1. | |
200 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction