Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hexamethylene bis-acetamide inducible 1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89007022 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-022 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-022 Supplier Abnova Corporation Supplier No. H00010614B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HEXIM1 protein.

Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells. This gene has no introns. [provided by RefSeq

Sequence: MAEPFLSEYQHQPQTSNCTGAAAVQEELNPERPPGAEERVPEEDSRWQSRAFPQLGGRPGPEGEGSLESQPPPLQTQACPESSCLREGEKGQNGDDSSAGGDFPPPAEVEPTPEAELLAQPCHDSEASKLGAPAAGGEEEWGQQQRQLGKKKHRRRPSKKKRHWKPYYKLTWEEKKKFDEKQSLRASRIRAEMFAKGQPVAPYNTTQFLMDDHDQEEPDLKTGLYSKRAAAKSDDTSDDDFMEEGGEEDGGSDGMGGDGSEFLQRDFSETYERYHTESLQNMSKQELIKEYLELEKCLSRMEDENNRLRLESKRLGGDDARVRELELELDRLRAENLQLLTENELHRQQERAPLSKFGD

Specifications

Antigen hexamethylene bis-acetamide inducible 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HEXIM1 protein.
Formulation No additive
Gene HEXIM1
Gene Accession No. BC006460
Gene Alias CLP1/EDG1/FLJ13562/HIS1/MAQ1
Gene Symbols HEXIM1
Host Species Mouse
Immunogen HEXIM1 (AAH06460, 1 a.a. ∼ 359 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 10614
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.