Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hey 1/HRT1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5714MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5714MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5714MI Supplier MilliporeSigma Supplier No. AB5714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Hey 1/HRT1 Antibody detects level of Hey 1/HRT1 and has been published and validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hey 1/HRT1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.
Gene Symbols HEY1; CHF-2; MGC1274; CHF2; HERP2; HRT1; hHRT1; HRT-1; HESR1; OAF1; HESR-1
Host Species Rabbit
Immunogen Synthetic peptide, amino acids 207-220 of human Hey 1 / HRT1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001040708.1; NM_012258.3
Target Species Human, Mouse
Content And Storage 2-8°C for up to 2 months, −70°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.