Learn More
Description
This gene encodes a nuclear protein belonging to the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcriptional repressors. Expression of this gene is induced by the Notch and c-Jun signal transduction pathways. Two similar and redundant genes in mouse are required for embryonic cardiovascular development, and are also implicated in neurogenesis and somitogenesis. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Specifications
Specifications
| Antigen | HEY1 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3E5 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant HEY1. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HEY1 |
| Gene Accession No. | NM_012258 |
| Gene Alias | BHLHb31/CHF2/HERP2/HESR1/HRT-1/MGC1274/OAF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.