Learn More
Description
This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined. [provided by RefSeq
Specifications
Specifications
| Antigen | HEYL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1E2 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant HEYL. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HEYL |
| Gene Accession No. | NM_014571 |
| Gene Alias | HRT3/MGC12623/bHLHb33 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.