Learn More
HEYL, Mouse, Clone: 2E12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HEYL.
Supplier: Abnova Corporation H00026508M22
Description
This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined. [provided by RefSeq
Sequence: MKRPKEPSGSDGESDGPIDVGQEGQLSQMARPLSTPSSSQMQARKKRRGIIEKRRRDRINSSLSELRRLVSpecifications
HEYL | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_014571 | |
HEYL | |
HEYL (NP_055386, 1 a.a. ∼ 70 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2E12 | |
Mouse monoclonal antibody raised against a partial recombinant HEYL. | |
HEYL | |
HRT3/MGC12623/bHLHb33 | |
Mouse | |
Affinity chromatography | |
RUO | |
26508 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.