Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HFE, Mouse, Clone: 1G12, Abnova™

Catalog No. 89022471 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-471 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-471 Supplier Abnova Corporation Supplier No. H00003077M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant HFE.

The protein encoded by this gene is a membrane protein that is similar to MHC class I-type proteins and associates with beta2-microglobulin (beta2M). It is thought that this protein functions to regulate iron absorption by regulating the interaction of the transferrin receptor with transferrin. The iron storage disorder, hereditary haemochromatosis, is a recessive genetic disorder that results from defects in this gene. At least nine alternatively spliced variants have been described for this gene. Additional variants have been found but their full-length nature has not been determined. [provided by RefSeq

Sequence: SHTLQVILGCEMQEDNSTEGYWKYGYDGQDHLEFCPDTLDWRAAEPRAWPTKLEWERHKIRARQNRAYLERDCPAQLQQLLELGRGVLDQQ

Specifications

Antigen HFE
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 1G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant HFE.
Formulation PBS with no preservative; pH 7.4
Gene HFE
Gene Accession No. NM_000410
Gene Alias HFE1/HH/HLA-H/MGC103790/dJ221C16.10.1
Gene Symbols HFE
Host Species Mouse
Immunogen HFE (NP_000401, 115 a.a. ∼ 205 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3077
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.