Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HHEX, Mouse, Clone: 4E9, Abnova™

Catalog No. 89022480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-480 Supplier Abnova Corporation Supplier No. H00003087M10
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant HHEX.

This gene encodes a member of the homeobox family of transcription factors, many of which are involved in developmental processes. Expression in specific hematopoietic lineages suggests that this protein may play a role in hematopoietic differentiation. [provided by RefSeq

Sequence: PLHKRKGGQVRFSNDQTIELEKKFETQKYLSPPERKRLAKMLQLSERQVKTWFQNRRAKWRRLKQENPQSNKKEELESLDSSCDQRQDLPSEQNKGASLDSSQCS

Specifications

Antigen HHEX
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4E9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant HHEX.
Formulation PBS with no preservative; pH 7.4
Gene HHEX
Gene Accession No. NM_002729
Gene Alias HEX/HMPH/HOX11L-PEN/PRH/PRHX
Gene Symbols HHEX
Host Species Mouse
Immunogen HHEX (NP_002720, 133 a.a. ∼ 237 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3087
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.