Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hHR23B Mouse, Unlabeled, Clone: 16, BD

Catalog No. BDB611018 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611018 50 μg
BDB611019 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611018 Supplier BD Biosciences Supplier No. 611018

Mouse Monoclonal Antibody

Nucleotide excision repair (NER) is an essential element of DNA repair which eliminates unrelated DNA base lesions, including those induced by UV irradiation or chemicals. Although well defined in prokaryotes, eukaryotic NER is complex and involves at least two distinct subpathways. The transcription-coupled repair subpathway eliminates DNA lesions just prior to transcription. This is important for the removal of lesions for which the second pathway, global genome repair (i.e. covers the entire genome), is unable to repair. Individuals with defects in both pathways may suffer from xeroderma pigmentosum (XP), a condition with increased incidence of cancer in skin and internal organs. Mutations in seven genes result in human XP. However, one of these, XP-C, affects only the global genome repair pathway. The XP-C protein interacts with hHR23B, the human homolog of RAD23, an essential component of yeast NER. The XPC-hHR23B complex is required for excision of thymine dimers. In addition, hHR23B contains an N-terminal ubiquitin-like domain that allows for proteasome interaction. Thus, hHR23B may be an essential component of NER and mechanisms of DNA repair.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen hHR23B
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 16
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human hHR23B aa. 73-193
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.