Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIC1, Mouse, Clone: 2B9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HIC1.
Supplier: Abnova Corporation H00003090M12
Description
Sequence: PEGVFAVARLTAEQLSLKQQDKAAAAELLAQTTHFLHDPKVALESLYPLAKFTAELGLSPDKAAEVLSQGAHLAAGPDGSpecifications
| HIC1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006497 | |
| HIC1 | |
| HIC1 (NP_006488.2, 627 a.a. ∼ 705 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2B9 | |
| Mouse monoclonal antibody raised against a partial recombinant HIC1. | |
| HIC1 | |
| ZBTB29/hic-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3090 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction