Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hic5 Mouse, Unlabeled, Clone: 34, BD

Catalog No. BDB611165 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611165 150 μg
BDB611164 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611165 Supplier BD Biosciences Supplier No. 611165
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Focal adhesions are cell structures terminal to the actin-stress fiber bundles. They attach cultured cells to their substratum or extracellular matrix. These adhesions occur through the integrin receptor molecules which link cytoskeletal proteins and extracellular matrix. The intracellular adhesion is composed of a number of proteins such as paxillin, VASP, vinculin, and the focal-adhesion kinases FAK and PYK2/CAKβ. Hic-5, also described as hydrogen peroxide inducible-mRNA, is a focal adhesion protein that binds to FAK and PYK2. It is
ubiquitously expressed with the highest levels found in lung, spleen, and heart. Induction of Hic-5 is accomplished by hydrogen peroxide or TGFβ1 and is repressed in K-ras transformed cells. Like paxillin, Hic-5 is tyrosine phosphorylated in Src-transformed cells and is highly similar to paxillin in its primary structure. Both proteins contain LIM domains and LD motifs. In addition, Hic-5 localizes to focal adhesions and co-immunoprecipitates with PYK2/CAKβ in vivo. Thus, Hic-5 may be a substrate for CAKβ and play a role in signal transduction during proliferation, cell motility, and adhesion.

Host Species: Mouse
Clone: 34
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse Hic-5 aa. 73-186
Formula Weight [Chemical]: 50kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hic5
Applications Western Blot
Classification Monoclonal
Clone 34
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Hic-5 aa. 73-186
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.