Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1 alpha Antibody - Exon 10 - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen HIF-1 alpha
Concentration 1.0 mg/mL
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP147179SS
SDP
View Documents
Novus Biologicals
NBP147179SS
0.025 mL
Each for $206.50
Only null left
Add to Cart
 
NBP147179
SDP
View Documents
Novus Biologicals
NBP147179
0.1 mL
Each for $426.00
Only null left
Add to Cart
 
Description

Description

HIF-1 alpha Polyclonal specifically detects HIF-1 alpha in Human samples. It is validated for Western Blot.
Specifications

Specifications

HIF-1 alpha
Western Blot 1.0 ug/ml
Polyclonal
Rabbit
Adaptive Immunity, Angiogenesis, Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Chromatin Research, Cytokine Research, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators
PBS and 30% Glycerol with 0.1% Sodium Azide
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hyp
HIF1A
IgG
Affinity Purified
HIF-1 alpha Antibody - Exon 10 is specific for Exon 10 of HIF-1 alpha.
1.0 mg/mL
Western Blot
Unconjugated
RUO
Human
Q16665
3091
This HIF-1 alpha Antibody - Exon 10 was developed against a synthetic peptide made to an internal portion of the human HIF-1 alpha protein (within amino acids 400 - 450) [Uniprot# Q16665].
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
93 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.