Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1β/ARNT1 Mouse, Unlabeled, Clone: 29, BD

Catalog No. BDB611079 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611079 150 μg
BDB611078 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB611079 Supplier BD Biosciences Supplier No. 611079
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Ah-receptor (AHR) is a ligand activated transcription factor that mediates the biological effects of agonists. AHR dimerizes with a structurally related protein known as ARNT (arylhydrocarbon-receptor nuclear transducer). This heterodimer binds enhancer elements and induces the expression of target genes, specifically those involved in the metabolism of xenobiotics. ARNT1 and ARNT2 are members of the basic-helix-loop-helix-PAS family of heterodimeric transcription factors, which also includes AHR, hypoxia-inducible factor-1α (HIF-1α), and the Drosophila single-minded protein (Sim). While ARNT2 expression is limited to brain and kidney, ARNT1 exhibits ubiquitous
expression. A targeted disruption of the Arnt locus in the mouse yields embryonic stem cells that fail to activate genes that normally respond to low oxygen tension. Arnt -/- embryos do not survive and show defective angiogenesis of the yolk sac and branchial arches, stunted development, and wasting. Thus, in addition to its regulation of xenobiotic metabolism genes, ARNT is thought to induce developmental gene expression resulting in vascularization of the developing embryo.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-1β/ARNT1
Applications Western Blot
Classification Monoclonal
Clone 29
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HIF-1β/ARNT1 aa. 461-574
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.