Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIF-1α Mouse anti-Human, Unlabeled, Clone: 54, BD

Catalog No. BDB610958 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610958 50 μg
BDB610959 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610958 Supplier BD Biosciences Supplier No. 610958
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Molecular oxygen (O2) is essential for mammalian metabolic processes such as oxidative phosphorylation. Thus, survival depends upon instantaneous transcriptional modulation of genes that maintain O2 homeostasis. Transcriptional control of several of these genes is mediated by hypoxia-inducible factor 1 (HIF-1). HIF-1 is a heterodimer whose α and β subunits are members of the PAS family of basic helix-loop-helix (bHLH) transcription factors. Common structural features of these proteins are an N-terminal bHLH DNA-binding domain and multiple PAS domains that confer dimerization ability and target gene specificity. Members diverge in their C-terminal regions. HIF-1β is also known as the arylhydrocarbon nuclear translocator which is part of the functional dioxin receptor. However, HIF-1α functions exclusively to mediate responses to O2 deprivation. It contains C-terminal and internal transactivation domains. Although HIF-1 aproteinlevels increase during hypoxia, it is unstable in the presence of O2 due to an oxygen-dependent degradation domain (ODD) that targets it for ubiquitination. Thus, HIF-1α is essential for functional HIF-1 to mediate gene transcription in response to hypoxia.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen HIF-1α
Applications Western Blot
Classification Monoclonal
Clone 54
Concentration 250μg/mL
Conjugate Unconjugated
Description MOP1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HIF-1α aa. 610-727
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.