Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIF3A Rabbit anti-Human, Mouse, DyLight™ 550, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522718
Description
Suggested positive control: reticulocyte lysate.
Hypoxia-inducible factor (HIF) is one of the most important factors in the cellular response to hypoxia, transcriptionally activating genes encoding proteins that mediate adaptive responses to reduced oxygen availability. HIF is a heterodimer consisting of one of three subunits, HIF1 alpha, HIF2 alpha, or HIF3 alpha. HIF target genes play critical roles in metabolism, angiogenesis, cell proliferation and cell survival. HIF3 alpha protein is one of several alpha/beta-subunit heterodimeric transcription factors that regulate many adaptive responses to low oxygen tension (hypoxia). The alpha 3 subunit lacks the transactivation domain found in factors containing either the alpha 1 or alpha 2 subunits. HIF3 alpha may be a marker for tumor growth and angiogenesis.
Specifications
HIF3A | |
Polyclonal | |
DyLight 550 | |
Q9Y2N7, Q0VBL6 | |
HIF3A | |
Bacterially expressed human HIF-3 alpha (N-terminus). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
HIF3A | |
HIF-3A, IPAS, MOP7, PASD7, bHLHe17, HIF3-alpha-1, PAS domain-containing protein 7, basic-helix-loop-helix-PAS protein MOP7, class E basic helix-loop-helix protein 17, hypoxia-inducible factor 3-alpha, inhibitory PAS domain protein, member of PAS protein 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53417, 64344 | |
4° C, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction