Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIPK2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA514471
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
HIPK2, a member of the KIPK subfamily of Ser/Thr protein kinases, phosphorylates homeodomain transcription factors. It may play a role as a corepressor for homeodomain transcription factors. This nuclear protein has been shown to interact with TRADD. It is highly expressed in neuronal tissues, heart and kidney, and weakly expressed in a ubiquitous way. HIPK2 is a target for sumoylation, and when conjugated it is directed to nuclear speckles
Specifications
HIPK2 | |
Polyclonal | |
HIPK2 | |
Homeodomain-interacting protein kinase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28996 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
Q9H2X6 | |
HIPK2 | |
KLH conjugated synthetic peptide between 344-372 amino acids from human HIPK2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction