Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIRA Mouse anti-Mouse, Human, Clone: WC119, MilliporeSigma™

Catalog No. 041488MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-148-8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-148-8MI Supplier MilliporeSigma Supplier No. 041488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-HIRA Antibody, clone WC119 is a Mouse Monoclonal Antibody for detection of HIRA also known as HIR (histone cell cycle regulation defective) homolog and has been validated in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIRA
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone WC119
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_003316.3
Gene Symbols HIRA, DGCR1, HIR, TUP1, TUPLE1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human HIRA.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.