Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIR histone cell cycle regulation defective homolog A (S. cerevisiae), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016925 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-925 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-925 Supplier Abnova Corporation Supplier No. H00007290B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human HIRA protein.

This gene encodes a histone chaperone that preferentially places the variant histone H3.3 in nucleosomes. Orthologs of this gene in yeast, flies, and plants are necessary for the formation of transcriptionally silent heterochomatin. This gene plays an important role in the formation of the senescence-associated heterochromatin foci. These foci likely mediate the irreversible cell cycle changes that occur in senescent cells. It is considered the primary candidate gene in some haploinsufficiency syndromes such as DiGeorge syndrome, and insufficient production of the gene may disrupt normal embryonic development. [provided by RefSeq

Sequence: MKLLKPTWVNHNGKPIFSVDIHPDGTKFATGGQGQDSGKVVIWNMSPVLQEDDEKDENIPKMLCQMDNHLACVNCVRWSNSGMYLASGGDDKLIMVWKRATYIGPSTVFGSSGKLANVEQWRCVSILRNHSGDVMDVAWSPHDAWLASCSVDNTVVIWNAVKFPEILATLRGHSGLVKGLTWDPVGKYIASQADDRSLKVWRTLDWQLETSITKPFDECGGTTHVLRLSWSPDGHYLVSAHAMNNSGPTAQIIEREGWKTNMDFVGHRKAVTVVKFNPKIFKKKQKNGSSAKPSCPYCCCAVGSKDRSLSVWLTCLKRPLVVIHELFDKSIMDISWTLNGLGILVCSMDGSVAFLDFSQDELGDPLSEEEKSRIHQSTYGKSLAIMTEAQLSTAVIENPEMLKYQRRQQQQQLDQKSAATREMGSATSVAGVVNGESLEDIRKNLLKKQVETRTADGRRRITPLCIAQLDTGDFSTAFFNSIPLSGSLAGTMLSSHSSPQLLPLDSSTPNSFGASKPCTEPVVAASARPAGDSVNKDSMNATSTPAALSPSVLTTPSKIEPMKAFDSRFTERSKATPGAPALTSMTPTAVERLKEQNLVKELRPRDLLESSSDSDEKVPLAKASSLSKRKLELEVETVEKKKKGRPRKDSRLMPVSLSVQSPAALTAEKEAMCLSAPALALKLPIPSPQRAFTLQVSSDPSMYIEVENEVTVVGGVKLSRLKCNREGKEWETVLTSRILTAAGSCDVVCVACEKRMLSVFSTCGRRLLSPILLPSPISTLHCTGSYVMALTAAATLSVWDVHRQVVVVKEESLHSILAGSDMTVSQILLTQHGIPVMNLSDGKAYCFNPSLSTWNLVSDKQDSLAQCADFRSSLPSQDAMLCSGPLAIIQGRTSNSGRQAARLFSVPHVVQQETTLAYLENQVAAALTLQSSHEYRHWLLVYARYLVNEGFEYRLREICKDLLGPVHYSTGSQWESTVVGLRKRELLKELLPVIGQNLRFQRLFTECQEQLDILRDK

Specifications

Antigen HIR histone cell cycle regulation defective homolog A (S. cerevisiae)
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human HIRA protein.
Formulation PBS with no preservative; pH 7.4
Gene HIRA
Gene Accession No. NM_003325.3
Gene Alias DGCR1/TUP1/TUPLE1
Gene Symbols HIRA
Host Species Mouse
Immunogen HIRA (NP_003316.3, 1 a.a. ∼ 1017 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 7290
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.