Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST2H4A, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89203577 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-203-577 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-203-577 Supplier Abnova Corporation Supplier No. H00008370K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human HIST2H4A peptide using ARM Technology.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in a histone cluster on chromosome 1. This gene is one of four histone genes in the cluster that are duplicated; this record represents the centromeric copy. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen HIST2H4A
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description histone cluster 2, H4a
Formulation In 1X PBS, pH 7.4
Gene HIST2H4A
Gene Accession No. HIST2H4A
Gene Alias FO108, H4, H4/n, H4F2, H4FN, HIST2H4
Gene Symbols HIST2H4A
Host Species Rabbit
Immunogen A synthetic peptide of human HIST2H4A
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8370
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.