Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Histidine Tagged Mouse anti-All, Clone: HIS.H8

Catalog No. 05949MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-949-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-949-MI Supplier MilliporeSigma™ Supplier No. 05949
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Histidine Tagged Antibody, clone HIS.H8 is validated for use in immunocytochemistry, immunoprecipitation, western blotting for the detection of Histidine Tagged.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histidine Tagged
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone HIS.H8
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2b in buffer containing PBS, pH 7.4 with 0.05% sodium azide.
Host Species Mouse
Immunogen 6x His synthetic peptide.
Purification Method Purified
Quantity 200 μg
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.