Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Mouse anti-Human, Bovine, Clone: F152C25W, MilliporeSigma™

Catalog No. MABE71MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE71MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE71MI Supplier MilliporeSigma Supplier No. MABE71
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Histone Antibody, Pan, clone F152.C25.WJJ is a Mouse Monoclonal Antibody for detection of Histone also known as Histone H1.4, Histone H1b and has been validated in western blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone
Applications ELISA, Western Blot
Classification Monoclonal
Clone F152C25W
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. N/A
Gene Symbols HIST1H1E, H1F4
Host Species Mouse
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.