Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Histone Deacetylase 1 (467-482), Polyclonal
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma™ PC54450UG
Description
Specifically detects Histone Deacetylase 1 (467-482) Clone: in Canine, Hamster, Human, Mouse, Rat samples, and it is validated for Immunofluorescence
The regulation of eukaryotic gene expression is marked by post-translational modification of nucleosomal histones, a key event that converts regions of chromosomes into transcriptionally active or inactive chromatin. A single nucleosomal core particle contains an octamer of histone proteins H2A, H2B, H3 and H4. Acetylated histone proteins confer accessibility of the DNA template to the transcriptional machinery. There are at least eight cloned human histone deacetylases. Studies have pointed to the possible involvement of histone deacetylases in human cancer. Histone Deacetylase 1 has been shown to interact directly with transcription factors and to deacetylate histone proteins H3 and H4. Studies have also shown that mammalian histone deacetylase (HDAC)-1, -2, and -3 are all capable of down-regulating p53 function. p53, the most commonly mutated gene in cancer cells, directs cell cycle arrest or induces programmed cell death (apoptosis) in response to stress. HDACs are classified by their sequence homology to the yeast HDACs. Class I proteins are related to Rpd3 and members of class II resemble HdaIp.
Specifications
Histone Deacetylase 1 (467-482) | |
Polyclonal | |
In PBS, 1mg/mL BSA. | |
a synthetic peptide corresponding to amino acids 467-482 of human histone deacetylase 1 | |
RUO | |
Recognizes the ∽62kDa HDAC1 protein in HeLa, HepG2, DU145, PC3, and MCF-7 cells. | |
−20°C | |
IgG |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
50 μg | |
Primary | |
Canine, Hamster, Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction