Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1° Mouse anti-Human, Mouse, Rat, Bovine, Xenopus, Clone: 34, MilliporeSigma™

Catalog No. MABE446MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE446MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE446MI Supplier MilliporeSigma Supplier No. MABE446
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Histone H1° Antibody, clone 34 is a highly specific mouse monoclonal antibody, that targets Histone H1 and has been tested in western blotting, immunocytochemistry, immunohistochemistry and Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H1°
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 34
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001069955
Gene Symbols H1F0
Host Species Mouse
Immunogen Recombinant protein corresponding to Ox liver Histone H1°
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat, Xenopus species
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.