Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Histone H1o/H5, Clone: 3H9,

Catalog No. 05629MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-629-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-629-MI Supplier MilliporeSigma™ Supplier No. 05629

Specifications

Antigen Histone H1o/H5
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3H9
Conjugate Unconjugated
Formulation PBS, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_005318
Gene Symbols H1F0, H1FV, MGC5241, H10, OTTHUMP00000028818
Host Species Mouse
Immunogen Native histone H5 protein purified from chicken erythrocytes; epitope mapped to amino acid residues 53-98.
Purification Method Affi-Gel blue purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.