Learn More
MilliporeSigma™ anti-Histone H2A (Acidic Patch), Polyclonal, ™ (Upstate™)
Supplier: MilliporeSigma™ 07146/DEL
Description
Histone H2A is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2A is involved with the structure of the nucleosomes of the «beads on a string« structure. GenBank Number: NM_003517. UniProt Number: Q16777 (Human). MW: 14kDa.
Chromatin Immunoprecipitation, Western Blotting
Specifications
Histone H2A (Acidic Patch) | |
Unconjugated | |
KLH-conjugated, synthetic peptide corresponding to amino acids 88-97 (IRNDEELNKL-C) of human Histone H2A | |
RUO | |
Primary | |
Serum |
Polyclonal | |
Rabbit | |
200 μL | |
Epigenetics | |
Chicken, Human, Mouse, Rat, Yeast | |