Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A.X Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07627MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-627-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-627-MI Supplier MilliporeSigma Supplier No. 07627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Histone H2A.X Antibody is a Rabbit Polyclonal Antibody for detection of Histone H2A.X also known as H2A histone family member X, H2AX histone and has been validated in PIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2A.X
Applications Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum containing 0.05% sodium azide and 30% glycerol. Liquid at −20°C.
Gene Symbols H2AFX; H2A/X; H2A.X; H2AX; H2a/x
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide (CSATVGPKAPSGGKKA) corresponding to amino acids 121-135 of human histone H2A.X, with an N-terminal cysteine added for conjugation purposes. The immunizing sequence has 13/15 identical amino acids in mouse.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_002096
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.