Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A.X Rabbit anti-Human, Mouse, Rat, Bovine, Polyclonal, MilliporeSigma™

Catalog No. AB10022MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10022MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10022MI Supplier MilliporeSigma Supplier No. AB10022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Histone H2A.X Antibody, Human (Rabbit Polyclonal Antibody) validated in western blotting to detect Histone H2A.X also known as H2AFX.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2A.X
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols H2AFX; H2AX
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human Histone H2A.X.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_002096
Target Species Bovine, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.