Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A.Z Sheep anti-Chicken, Human, Zebrafish, Rabbit, Rat, Xenopus, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09862MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-862-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-862-MI Supplier MilliporeSigma Supplier No. 09862

Sheep Polyclonal Antibody

Use Anti-Histone H2A.Z Antibody (Sheep Polyclonal Antibody) validated in western blotting to detect Histone H2A.Z also known as H2A histone family member Z, H2AZ histone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2A.Z
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified sheep polyclonal IgG in PBS, pH 7.4 containing with 1% BSA with 0.02% sodium azide.
Gene Symbols H2A.z; H2A/z ; H2AZ; MGC117173; H2AFZ
Host Species Sheep
Immunogen The immunogen was a linear peptide corresponding to C-terminal amino acids 113-127 of human Histone H2A.Z.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_036544
Target Species Chicken, Human, Mouse, Rabbit, Rat, Xenopus species, Zebrafish
Content And Storage 2-8°C for 12 months in undiluted aliquotes. Avoid freeze/thaw cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.