Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A.Z Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07594MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-594-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-594-MI Supplier MilliporeSigma Supplier No. 07594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Histone H2A.Z also known as H2AZ histone with Anti-Histone H2A.Z Antibody (Rabbit Polyclonal Antibody), that has been demonstrated to work in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2A.Z
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols H2AFZ; H2A/z; H2A.Z; MGC117173; H2AZ; H2A.z
Host Species Rabbit
Immunogen peptide containing the sequence SLIGKKGQQ, corres-ponding to the C-terminus of human histone H2A.Z
Purification Method Serum
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.