Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B Mouse anti-Mouse, Human, Rat, Clone: 5HH2-2A8, MilliporeSigma™

Catalog No. 051352MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-135-2MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-135-2MI Supplier MilliporeSigma Supplier No. 051352
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Histone H2B Antibody, clone 5HH2-2A8 is a Mouse Monoclonal Antibody for detection of Histone H2B also known as Histone H2B, histone 2 H2be and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2B
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5HH2-2A8
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1κ ascites with 0.05% sodium azide.
Gene Accession No. BC067487
Gene Symbols H2B, GL105, H2B-GL105, H2B.1, H2B.q, H2B/q, H2BFQ
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic linear peptide corresponding to the C-terminus of Histone H2B.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.