Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Histone H2B type 2-E (H2be), Polyclonal

Catalog No. ABE1384MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1384MI 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1384MI Supplier MilliporeSigma™ Supplier No. ABE1384
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Histone H2B type 2-E (H2be) Clone: in Human, Mouse, Rat samples, and it is validated for Dot Blot, Western Blotting

Histone H2B type 2-E (UniProt Q64524; also known as H2b 613, Histone 2 H2be) is encoded by the Hist2h2be gene (Gene ID 319190) in murine species. H2be is a replication-independent histone H2b variant expressed exclusively in olfactory chemosensory neurons. Levels of H2be are heterogeneous among olfactory neurons, but stereotyped according to the identity of the co-expressed olfactory receptor (OR). H2be is nearly undetectable in highly active neurons, but almost completely replaces H2b in inactive neurons. Changes in H2be expression affect olfactory function and OR representation in the adult olfactory epithelium. Genetic studies show that the deletion of H2be significantly extends the lifespan of neurons, whereas elevated H2be shortens their lifespan.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2B type 2-E (H2be)
Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q64524
Gene Symbols Hist2h2be
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminal sequence of mouse histone H2B type 2-E (H2be).
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Epigenetics, Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_835586
Test Specificity The identity of an immunoreactive band of ∽16kDa detected by this polyclonal antibody using HeLa cell lysate has not been determined, but most likely corresponds to human histone H2B type 1-L (Histone H2B.c; Uniprot Q99880) whose N-terminal sequence (up to a.a. 39) is identical to that of murine histone H2B type 2-E.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.