Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3.3 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09838MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-838-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-838-MI Supplier MilliporeSigma Supplier No. 09838
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Histone H3.3 Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry, DB, Chimmunoprecipitation to detect Histone H3.3 also known as Histone H3.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3.3
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols H3F3A; H3.3A; H3F3; PP781; H3F3B; H3.3B
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3.3.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_002098
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.