Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Rabbit anti-Chicken, Human, Mouse, Rat, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™

Catalog No. 07690MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-690-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-690-MI Supplier MilliporeSigma Supplier No. 07690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-Histone H3 Antibody, CT, pan (rabbit polyclonal antibody) validated in Chimmunoprecipitation, western blotting to detect Histone H3 also known as H3, Histone H3, H3 histone family member T, histone 3 H3, histone cluster 3 H3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3
Applications ChIP Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit antiserum diluted 1:2 in storage buffer (0.02M Phosphate, 0.25M NaCl, 0.1% sodium azide) before the addition of glycerol to 30%.
Gene Symbols HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to the C-terminus of human Histone H3.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002107.3
Target Species Chicken, Human, Mouse, Rat, S. cerevisiae, Yeast
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.