Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Mouse anti-Vertebrates, Green Plants, Human, Clone: 6.6.2, MilliporeSigma™

Catalog No. 05499MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-499-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-499-MI Supplier MilliporeSigma Supplier No. 05499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Histone H3 Antibody, clone 6.6.2 is a Mouse Monoclonal Antibody for detection of Histone H3 also known as H3 histone family 3A and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3
Applications Western Blot
Classification Monoclonal
Clone 6.6.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 70% storage buffer 0.1MTris-glycine, pH 7.4, 0.15MNaCl, with 0.05% sodium azide and 30% glycerol.
Gene Accession No. NP_003484
Gene Symbols H3F3A, H3F3, MGC87782, H3.3B, MGC87783, H3F3B, H3.3A
Host Species Mouse
Immunogen KLH conjugated peptide corresponding to amino acids 1-20 [ARTKQTARKSTGGKAPRKQLC] of human histone H3. Clone 6.6.2.
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Plant, Vertebrates
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.