Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Rabbit anti-Chicken, Human, Mouse, Rat, Yeast (S. cerevisiae), Clone: A3S, MilliporeSigma™

Catalog No. 04928MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-928-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-928-MI Supplier MilliporeSigma Supplier No. 04928
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-Histone H3 Antibody, CT, pan, clone A3S is a Rabbit Monoclonal Antibody for detection of Histone H3 also known as Histone H3 and has been validated in Chimmunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3
Applications ChIP Assay, Western Blot
Classification Monoclonal
Clone A3S
Conjugate Unconjugated
Formulation Cultured supernantant in 0.05% sodium azide
Gene Accession No. NM_003493
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to the C-terminus of human Histone H3
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) H3F3A, H3F3B
Target Species Chicken, Human, Mouse, Rat, S. cerevisiae, Yeast
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.