Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3 Mouse anti-Human, Vertebrates, Clone: CMA301, MilliporeSigma™
Mouse Monoclonal Antibody
$153.22 - $692.64
Specifications
| Antigen | Histone H3 |
|---|---|
| Clone | CMA301 |
| Applications | Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
05-134-1SMI
|
MilliporeSigma
051341S |
10 μg |
Each for $153.22
|
|
|||||
|
05-134-1MI
|
MilliporeSigma
051341 |
100 μg |
Each for $692.64
|
|
|||||
Description
Use Anti-Histone H3 Antibody, (Unmodified Lys4), clone CMA301, (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry, ELISA, immunoprecipitation, flow cytometry, DB to detect Histone H3 also known as histone 3-H3.Specifications
| Histone H3 | |
| Dot Blot, ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Human, Vertebrates | |
| NM_003493 | |
| Synthetic peptide corresponding to amino acids 1-12 of human Histone H3, conjugated to KLH. | |
| Primary | |
| 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
| CMA301 | |
| Monoclonal | |
| Mouse | |
| Mouse monoclonal IgG2bκ supernatant in PBS with 0.05% sodium azide. | |
| H3.4, H3/g , H3/t, H3FT, H3T, H3t, MGC126886, MGC126888, OTTHUMP00000037945 | |
| IgG2b κ | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title